Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DEPDC5
Entrez Gene ID
9681
Gene Name
DEP domain containing 5
Gene Aliases
DEP.5, FFEVF
Chromosome Location
Chr.22: 31753951 - 31907034 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136029.2 NP_001129501.1 38 - 39 4240 88
NM_001242896.1 NP_001229825.1 39 - 40 4409 88
NM_001242897.1 NP_001229826.1 36 - 37 4013 88
NM_014662.4 NP_055477.1 37 - 38 4174 88
NR_110988.1 35 - 36 4000 88
XM_005261862.1 XP_005261919.1 39 - 40 4313 88
XM_011530557.2 XP_011528859.1 39 - 40 4389 88
XM_011530558.2 XP_011528860.1 38 - 39 4350 88
XM_011530559.2 XP_011528861.1 38 - 39 4323 88
XM_011530560.2 XP_011528862.1 37 - 38 4201 88
XM_011530561.2 XP_011528863.1 37 - 38 4155 88
XM_011530563.2 XP_011528865.1 36 - 37 4116 88
XM_011530569.2 XP_011528871.1 16 - 17 2180 88
XM_017029109.1 XP_016884598.1 39 - 40 4286 88
XM_017029110.1 XP_016884599.1 38 - 39 4247 88
XM_017029111.1 XP_016884600.1 38 - 39 4220 88
XM_017029112.1 XP_016884601.1 37 - 38 4079 88
GenBank mRNA AB014545.1 - 37 - 38 4173 88
AJ698950.1 - 38 - 39 4235 88
AJ698951.1 - 36 - 37 3962 88
AJ704764.1 - 37 - 38 4169 88
AK294583.1 - 8 - 9 1146 88
AK294987.1 - 36 - 37 3985 88
AK309008.1 - 10 - 11 1222 88
AK316364.1 - 8 - 9 903 88
BC136612.1 - 39 - 40 4323 88
BC146766.1 - 37 - 38 4173 88
BX640828.1 - 15 - 16 2154 88
CR749304.1 - 17 - 18 2333 88

View Full Product Details