The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015370.1 | NP_056185.1 | 1 - 2 | 136 | 97 |
GenBank mRNA | AL035364.1 | - | 1 - 2 | 136 | 97 |
BC074758.2 | - | 1 - 2 | 54 | 97 | |
BC126262.1 | - | 1 - 2 | 58 | 97 | |
BC126264.1 | - | 1 - 2 | 58 | 97 |