Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRDM2
Entrez Gene ID
7799
Gene Name
PR domain 2
Gene Aliases
HUMHOXY1, KMT8, MTB-ZF, RIZ, RIZ1, RIZ2
Chromosome Location
Chr.1: 13698875 - 13825079 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007257.2 NP_001007258.1 3 - 4 4629 145
NM_015866.4 NP_056950.2 8 - 9 5889 145
XM_017002254.1 XP_016857743.1 7 - 8 5111 145
XM_017002257.1 XP_016857746.1 7 - 8 5119 145
XM_017002262.1 XP_016857751.1 4 - 5 4650 145
GenBank mRNA BX647310.1 - 4 - 5 4652 145
D45132.1 - 3 - 4 4610 145

View Full Product Details