The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016370.2 | NP_057454.1 | 1 - 2 | 221 | 106 |
GenBank mRNA | AB036693.1 | - | 1 - 2 | 167 | 106 |
AK313425.1 | - | 1 - 2 | 116 | 106 | |
DA501604.1 | - | 1 - 2 | 221 | 107 |