Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEPSECS
Entrez Gene ID
51091
Gene Name
Sep (O-phosphoserine) tRNA:Sec (selenocysteine) tRNA synthase
Gene Aliases
LP, PCH2D, SLA, SLA/LP
Chromosome Location
Chr.4: 25120005 - 25160582 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF146396.2 - 1 - 2 33 61
BC023539.2 - - 177 61

View Full Product Details