The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017518.7 | NP_059988.3 | 4 - 5 | 429 | 75 |
GenBank mRNA | AF267739.1 | - | 4 - 5 | 360 | 75 |
AK296147.1 | - | 7 - 8 | 744 | 75 | |
AK300650.1 | - | 4 - 5 | 381 | 75 | |
AK302066.1 | - | 4 - 5 | 360 | 75 | |
BC008141.1 | - | 4 - 5 | 429 | 75 |