The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001316943.1 | NP_001303872.1 | 5 - 6 | 1255 | 93 |
NM_001316944.1 | NP_001303873.1 | 3 - 4 | 910 | 93 | |
XM_006720590.2 | XP_006720653.1 | 5 - 6 | 1296 | 93 | |
XM_017022386.1 | XP_016877875.1 | 5 - 6 | 1239 | 93 | |
XM_017022387.1 | XP_016877876.1 | 4 - 5 | 1160 | 93 | |
XM_017022389.1 | XP_016877878.1 | 4 - 5 | 1089 | 93 | |
GenBank mRNA | AK000516.1 | - | 3 - 4 | 910 | 93 |
AK058010.1 | - | 5 - 6 | 839 | 93 |