The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289067.1 | NP_001275996.1 | 19 - 20 | 2381 | 93 |
NM_001289068.1 | NP_001275997.1 | 18 - 19 | 2198 | 93 | |
NM_001289069.1 | NP_001275998.1 | 18 - 19 | 2147 | 93 | |
NM_001289070.1 | NP_001275999.1 | 16 - 17 | 1949 | 93 | |
NM_001289071.1 | NP_001276000.1 | 19 - 20 | 2287 | 93 | |
NM_001289072.1 | NP_001276001.1 | 16 - 17 | 1853 | 93 | |
NM_001289073.1 | NP_001276002.1 | 17 - 18 | 2178 | 93 | |
NM_001289074.1 | NP_001276003.1 | 19 - 20 | 2162 | 93 | |
NM_001289075.1 | NP_001276004.1 | 17 - 18 | 2046 | 93 | |
NM_018063.4 | NP_060533.2 | 18 - 19 | 2243 | 93 | |
GenBank mRNA | AB102716.1 | - | 19 - 20 | 2132 | 93 |
AB102717.1 | - | 17 - 18 | 2023 | 93 | |
AB102718.1 | - | 18 - 19 | 1992 | 93 | |
AB102719.1 | - | 16 - 17 | 1698 | 93 | |
AB102720.1 | - | 17 - 18 | 1891 | 93 | |
AB102721.1 | - | 16 - 17 | 1794 | 93 | |
AB102722.1 | - | 19 - 20 | 2007 | 93 | |
AB102723.1 | - | 19 - 20 | 2226 | 93 | |
AB113249.1 | - | 18 - 19 | 2088 | 93 | |
AF155827.1 | - | 18 - 19 | 2193 | 93 | |
AK001201.1 | - | 4 - 5 | 401 | 93 | |
AK314485.1 | - | 18 - 19 | 2232 | 93 | |
AY007108.1 | - | 6 - 7 | 700 | 93 | |
BC015477.1 | - | 5 - 6 | 526 | 93 | |
BC030963.1 | - | 7 - 8 | 846 | 93 | |
BC031004.1 | - | 7 - 8 | 846 | 93 | |
BX538033.1 | - | 18 - 19 | 2182 | 93 |