Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 56452522-56453021 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASS4
Entrez Gene ID
57091
Gene Name
Cas scaffolding protein family member 4
Gene Aliases
C20orf32, CAS4, HEFL, HEPL
UniGene
Hs.473144
Gene Chromosome Location
Chr.20: 56411550-56460240 on Build GRCh38
Transcripts
NM_001164114.1 NM_001164115.1 NM_001164116.1 NM_020356.3 XM_006723831.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1028285 c.1829A>C p.K610T Chr.20: 56453005-56453005 on Build GRCh38
2764001 c.1619G>A p.R540H Chr.20: 56452795-56452795 on Build GRCh38
3548045 c.1603G>A p.E535K Chr.20: 56452779-56452779 on Build GRCh38
1028281 c.1444T>C p.S482P Chr.20: 56452620-56452620 on Build GRCh38
1412631 c.1493G>A p.R498Q Chr.20: 56452669-56452669 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771080322 Chr.20: 56452668-56452668 on Build GRCh38
rs772325603 Chr.20: 56452889-56452889 on Build GRCh38
rs747621928 Chr.20: 56452619-56452619 on Build GRCh38
rs749307548 Chr.20: 56452665-56452665 on Build GRCh38
rs148921681 Chr.20: 56452759-56452759 on Build GRCh38
rs200192753 Chr.20: 56452807-56452807 on Build GRCh38
rs764139797 Chr.20: 56452722-56452722 on Build GRCh38
rs774042931 Chr.20: 56452933-56452933 on Build GRCh38
rs757429813 Chr.20: 56452876-56452876 on Build GRCh38
rs199513148 Chr.20: 56452944-56452944 on Build GRCh38
rs749857293 Chr.20: 56452592-56452592 on Build GRCh38
rs757585031 Chr.20: 56452969-56452969 on Build GRCh38
rs917517655 Chr.20: 56452608-56452608 on Build GRCh38
rs550251334 Chr.20: 56452735-56452735 on Build GRCh38
rs768670405 Chr.20: 56452793-56452793 on Build GRCh38
rs564505566 Chr.20: 56452724-56452724 on Build GRCh38
rs762687571 Chr.20: 56452841-56452841 on Build GRCh38
rs142811598 Chr.20: 56452838-56452838 on Build GRCh38
rs533336238 Chr.20: 56452725-56452725 on Build GRCh38
rs904655774 Chr.20: 56452696-56452696 on Build GRCh38
rs1044229460 Chr.20: 56452587-56452587 on Build GRCh38
rs557015028 Chr.20: 56452924-56452924 on Build GRCh38
rs757945426 Chr.20: 56452601-56452601 on Build GRCh38
rs146122261 Chr.20: 56452603-56452603 on Build GRCh38
rs950832531 Chr.20: 56452932-56452932 on Build GRCh38
rs536166322 Chr.20: 56452814-56452814 on Build GRCh38
rs527807357 Chr.20: 56452548-56452548 on Build GRCh38
rs1029721066 Chr.20: 56452751-56452751 on Build GRCh38
rs770573097 Chr.20: 56452931-56452931 on Build GRCh38
rs758545822 Chr.20: 56452996-56452996 on Build GRCh38
rs761675318 Chr.20: 56452795-56452795 on Build GRCh38
rs778992928 Chr.20: 56452781-56452781 on Build GRCh38
rs776832429 Chr.20: 56452543-56452543 on Build GRCh38
rs755415780 Chr.20: 56452756-56452756 on Build GRCh38
rs775960291 Chr.20: 56452945-56452945 on Build GRCh38
rs770317129 Chr.20: 56452644-56452644 on Build GRCh38
rs201366703 Chr.20: 56452864-56452864 on Build GRCh38
rs759018577 Chr.20: 56452934-56452934 on Build GRCh38
rs529524026 Chr.20: 56452758-56452758 on Build GRCh38
rs916964194 Chr.20: 56452892-56452892 on Build GRCh38
rs377729881 Chr.20: 56452740-56452740 on Build GRCh38
rs749589898 Chr.20: 56452772-56452772 on Build GRCh38
rs767310064 Chr.20: 56452564-56452564 on Build GRCh38
rs144538897 Chr.20: 56452562-56452562 on Build GRCh38
rs764510496 Chr.20: 56452955-56452955 on Build GRCh38
rs919291093 Chr.20: 56452565-56452565 on Build GRCh38
rs753526844 Chr.20: 56452582-56452582 on Build GRCh38
rs1044586660 Chr.20: 56452660-56452660 on Build GRCh38
rs891477283 Chr.20: 56452726-56452726 on Build GRCh38
rs761184541 Chr.20: 56452742-56452742 on Build GRCh38
rs762367653 Chr.20: 56452729-56452729 on Build GRCh38
rs767993594 Chr.20: 56452850-56452850 on Build GRCh38
rs1016759262 Chr.20: 56452800-56452800 on Build GRCh38
rs760752577 Chr.20: 56452717-56452717 on Build GRCh38
rs755326072 Chr.20: 56452641-56452641 on Build GRCh38
rs151045039 Chr.20: 56452914-56452914 on Build GRCh38
rs962517223 Chr.20: 56452804-56452804 on Build GRCh38
rs992861360 Chr.20: 56452997-56452997 on Build GRCh38
rs201454304 Chr.20: 56452659-56452659 on Build GRCh38
rs778255229 Chr.20: 56452866-56452866 on Build GRCh38
rs775284457 Chr.20: 56452669-56452669 on Build GRCh38
rs781242436 Chr.20: 56452905-56452905 on Build GRCh38
rs765956674 Chr.20: 56452822-56452822 on Build GRCh38
rs756912515 Chr.20: 56452586-56452586 on Build GRCh38
rs371141970 Chr.20: 56452825-56452825 on Build GRCh38
rs762611920 Chr.20: 56452818-56452818 on Build GRCh38
rs753246501 Chr.20: 56452860-56452860 on Build GRCh38
rs745893482 Chr.20: 56452884-56452884 on Build GRCh38
rs189999295 Chr.20: 56452908-56452908 on Build GRCh38
rs764045834 Chr.20: 56452568-56452568 on Build GRCh38
rs754186572 Chr.20: 56452962-56452962 on Build GRCh38
rs371150425 Chr.20: 56452728-56452728 on Build GRCh38
rs764981924 Chr.20: 56452591-56452591 on Build GRCh38
rs536126976 Chr.20: 56452998-56452998 on Build GRCh38
rs770021620 Chr.20: 56452559-56452559 on Build GRCh38
rs765534010 Chr.20: 56452970-56452970 on Build GRCh38
rs1028497835 Chr.20: 56452798-56452798 on Build GRCh38
rs777828565 Chr.20: 56452766-56452766 on Build GRCh38
rs16979936 Chr.20: 56452648-56452648 on Build GRCh38
rs145762733 Chr.20: 56452651-56452651 on Build GRCh38
rs186338077 Chr.20: 56452794-56452794 on Build GRCh38
rs750712364 Chr.20: 56452988-56452988 on Build GRCh38
rs866071361 Chr.20: 56452650-56452650 on Build GRCh38
rs140151545 Chr.20: 56452622-56452622 on Build GRCh38
rs779035230 Chr.20: 56452879-56452879 on Build GRCh38
rs774946832 Chr.20: 56452787-56452787 on Build GRCh38
rs376814976 Chr.20: 56452566-56452566 on Build GRCh38
rs559542055 Chr.20: 56452542-56452542 on Build GRCh38
rs770901628 Chr.20: 56452779-56452779 on Build GRCh38
rs773665975 Chr.20: 56452967-56452968 on Build GRCh38
rs761183534 Chr.20: 56452946-56452946 on Build GRCh38
rs571184962 Chr.20: 56452869-56452869 on Build GRCh38
rs992522298 Chr.20: 56452880-56452880 on Build GRCh38
rs534428950 Chr.20: 56452809-56452809 on Build GRCh38
rs573881231 Chr.20: 56452925-56452925 on Build GRCh38
rs138464146 Chr.20: 56452694-56452694 on Build GRCh38

View Full Product Details