The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|---|
RefSeq | NM_001281482.1 | NP_001268411.1 | 7 - 8 | 768 | 63 | |
NM_001281483.1 | NP_001268412.1 | 6 - 7 | 658 | 63 | ||
NM_001281484.1 | NP_001268413.1 | 6 - 7 | 667 | 63 | ||
NM_018209.3 | NP_060679.1 | 7 - 8 | 768 | 63 | ||
NM_175609.2 | NP_783202.1 | 7 - 8 | 768 | 63 | ||
XM_006723823.2 | XP_006723886.1 | 7 - 8 | 751 | 63 | ||
XM_011528901.1 | XP_011527203.1 | 7 - 8 | 751 | 63 | ||
XM_017027950.1 | XP_016883439.1 | 7 - 8 | 834 | 63 | ||
GenBank mRNA | AK001629.1 | - | 7 - 8 | 740 | 63 | |
AK027268.1 | - | 4 - 5 | 460 | 63 | ||
AK027441.1 | - | 2 - 3 | 163 | 63 | ||
AK223427.1 | - | 7 - 8 | 689 | 63 | ||
AK296351.1 | - | 6 - 7 | 630 | 63 | ||
AK303454.1 | - | 6 - 7 | 636 | 63 | ||
AK308161.1 | - | 7 - 8 | 751 | 63 | ||
AY927573.1 | - | LIT3574 AY927573 |
1 - 2 | 62 | 63 | |
BC000786.1 | - | 7 - 8 | 728 | 63 | ||
BC006085.1 | - | 7 - 8 | 719 | 63 | ||
BC011876.1 | - | 7 - 8 | 728 | 63 | ||
BC028233.1 | - | 7 - 8 | 688 | 63 |