The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031710.2 | NP_001026880.2 | 1 - 2 | 414 | 103 |
NM_001172428.1 | NP_001165899.1 | 1 - 2 | 414 | 103 | |
XM_006715753.2 | XP_006715816.1 | 1 - 2 | 409 | 103 | |
XM_006715757.3 | XP_006715820.1 | 1 - 2 | 409 | 103 | |
XM_017012440.1 | XP_016867929.1 | 1 - 2 | 409 | 103 | |
GenBank mRNA | AF111113.1 | - | 1 - 2 | 298 | 103 |
AK290479.1 | - | 1 - 2 | 208 | 103 | |
AK297369.1 | - | - | 339 | 103 | |
AK297421.1 | - | 1 - 2 | 415 | 103 | |
AL136597.1 | - | 1 - 2 | 360 | 103 | |
BC039585.1 | - | 1 - 2 | 356 | 103 | |
DC370850.1 | - | 1 - 2 | 414 | 103 |