The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184993.1 | NP_001171922.1 | 34 - 35 | 4802 | 121 |
NM_031277.2 | NP_112567.2 | 34 - 35 | 4814 | 121 | |
XM_006719846.3 | XP_006719909.1 | 35 - 36 | 5255 | 121 | |
XM_006719849.2 | XP_006719912.1 | 35 - 36 | 4972 | 121 | |
XM_006719852.2 | XP_006719915.1 | 34 - 35 | 4850 | 121 | |
XM_011535152.2 | XP_011533454.1 | 39 - 40 | 6880 | 121 | |
XM_011535155.2 | XP_011533457.1 | 38 - 39 | 5616 | 121 | |
XM_011535156.2 | XP_011533458.1 | 39 - 40 | 6880 | 121 | |
XM_011535157.2 | XP_011533459.1 | 39 - 40 | 6868 | 121 | |
XM_011535158.2 | XP_011533460.1 | 38 - 39 | 6754 | 121 | |
XM_011535159.2 | XP_011533461.1 | 38 - 39 | 6733 | 121 | |
XM_011535160.2 | XP_011533462.1 | 38 - 39 | 6721 | 121 | |
XM_011535162.1 | XP_011533464.1 | 31 - 32 | 4496 | 121 | |
XM_011535163.1 | XP_011533465.1 | 26 - 27 | 3766 | 121 | |
XM_011535164.2 | XP_011533466.1 | 20 - 21 | 2662 | 121 | |
XM_017020676.1 | XP_016876165.1 | 35 - 36 | 4924 | 121 | |
GenBank mRNA | AK001907.1 | - | 14 - 15 | 1909 | 121 |
AK302435.1 | - | 34 - 35 | 4778 | 121 | |
BC064847.1 | - | 18 - 19 | 2465 | 121 | |
BC146822.1 | - | 35 - 36 | 4792 | 121 | |
DB517763.1 | - | 1 - 2 | 153 | 121 |