The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193288.2 | NP_001180217.1 | 5 - 6 | 1452 | 98 |
NM_020344.3 | NP_065077.1 | 6 - 7 | 1503 | 98 | |
XM_005251425.2 | XP_005251482.1 | 7 - 8 | 1782 | 98 | |
XM_005251426.4 | XP_005251483.1 | 7 - 8 | 2314 | 98 | |
XM_006716750.3 | XP_006716813.1 | 6 - 7 | 2263 | 98 | |
XM_017014592.1 | XP_016870081.1 | 11 - 12 | 2253 | 98 | |
XM_017014593.1 | XP_016870082.1 | 6 - 7 | 1735 | 98 | |
GenBank mRNA | AF097366.1 | - | 6 - 7 | 1412 | 98 |
AF177987.1 | - | 5 - 6 | 1361 | 98 | |
BC069622.1 | - | 6 - 7 | 1400 | 98 | |
BC143889.1 | - | 5 - 6 | 1322 | 98 |