The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167576.1 | NP_001161048.1 | 4 - 5 | 1458 | 55 |
NM_001167577.1 | NP_001161049.1 | 5 - 6 | 1623 | 55 | |
NM_020389.2 | NP_065122.1 | 6 - 7 | 1806 | 55 | |
GenBank mRNA | AJ272034.1 | - | 5 - 6 | 1584 | 55 |
AJ549088.1 | - | 4 - 5 | 1401 | 55 | |
AJ549089.1 | - | 4 - 5 | 1419 | 55 | |
AJ549090.1 | - | 2 - 3 | 1019 | 55 | |
AY167926.1 | - | 4 - 5 | 1416 | 55 | |
AY167927.1 | - | 3 - 4 | 1236 | 55 | |
BC128185.1 | - | 5 - 6 | 1584 | 55 | |
BC128186.1 | - | 5 - 6 | 1600 | 55 |