The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171132.2 | NP_001164603.1 | 4 - 5 | 527 | 79 |
NM_001171134.2 | NP_001164605.1 | 3 - 4 | 624 | 79 | |
NM_001282311.1 | NP_001269240.1 | 4 - 5 | 722 | 177 | |
NM_001282312.1 | NP_001269241.1 | 4 - 5 | 468 | 79 | |
NM_021806.3 | NP_068578.2 | 3 - 4 | 624 | 79 | |
XM_005274714.3 | XP_005274771.1 | 5 - 6 | 572 | 177 | |
XM_005274716.3 | XP_005274773.1 | 5 - 6 | 572 | 177 | |
XM_005277879.4 | XP_005277936.1 | 4 - 5 | 533 | 79 | |
XM_006724831.2 | XP_006724894.1 | 5 - 6 | 572 | 177 | |
XM_006724832.3 | XP_006724895.1 | 4 - 5 | 533 | 79 | |
XM_006724833.2 | XP_006724896.1 | 5 - 6 | 572 | 177 | |
GenBank mRNA | AK024601.1 | - | 4 - 5 | 484 | 79 |
AK294116.1 | - | 3 - 4 | 587 | 79 | |
AK314682.1 | - | 3 - 4 | 542 | 79 | |
AL539139.3 | - | 4 - 5 | 719 | 177 | |
BC002934.1 | - | 4 - 5 | 626 | 177 | |
BC008912.2 | - | 3 - 4 | 275 | 79 | |
BM457676.1 | - | 4 - 5 | 509 | 79 | |
X87193.1 | - | 3 - 4 | 431 | 79 |