Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SH2D4A
Entrez Gene ID
63898
Gene Name
SH2 domain containing 4A
Gene Aliases
PPP1R38, SH2A
Chromosome Location
Chr.8: 19313443 - 19396218 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001174159.1 NP_001167630.1 6 - 7 1012 64
NM_001174160.1 NP_001167631.1 5 - 6 777 64
NM_022071.3 NP_071354.2 6 - 7 1162 64
XM_011544610.1 XP_011542912.1 5 - 6 1208 64
XM_017013729.1 XP_016869218.1 5 - 6 930 64
GenBank mRNA AK024620.1 - 4 - 5 431 64
AK024799.1 - 6 - 7 1040 64
AK225052.1 - 4 - 5 431 64
AK293301.1 - 5 - 6 653 64
BC014525.2 - 3 - 4 358 64
BC067117.1 - 3 - 4 372 64
BC082982.1 - 6 - 7 1012 64

View Full Product Details