Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- MLST8
- Entrez Gene ID
-
64223
- Gene Name
- MTOR associated protein, LST8 homolog
- Gene Aliases
-
GBL, GbetaL, LST8, POP3, WAT1
- Chromosome Location
-
Chr.16: 2205177 - 2209417 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK022227.1 |
- |
6 - 7 |
744 |
106 |
View Full Product Details