Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC25A23
Entrez Gene ID
79085
Gene Name
solute carrier family 25 member 23
Gene Aliases
APC2, MCSC2, SCaMC-3
Chromosome Location
Chr.19: 6436081 - 6460221 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024103.2 NP_077008.2 8 - 9 1216 86
XM_011528274.1 XP_011526576.1 9 - 10 1808 86
XM_011528275.1 XP_011526577.1 8 - 9 1667 86
XM_011528277.1 XP_011526579.1 9 - 10 1808 86
XM_011528278.1 XP_011526580.1 9 - 10 1808 86
XM_011528280.1 XP_011526582.1 9 - 10 1808 86
XM_011528281.1 XP_011526583.1 9 - 10 1808 86
XM_011528282.1 XP_011526584.1 9 - 10 1808 86
XM_011528283.1 XP_011526585.1 9 - 10 1808 86
XM_011528284.1 XP_011526586.1 9 - 10 1808 86
XM_017027285.1 XP_016882774.1 9 - 10 1808 86
XM_017027288.1 XP_016882777.1 8 - 9 1667 86
GenBank mRNA AJ512835.1 - 8 - 9 1181 86
AJ619962.1 - 8 - 9 1171 86
AJ619988.1 - 8 - 9 1181 86
AK054901.1 - 7 - 8 913 86
AK074579.1 - 3 - 4 338 86
AK294514.1 - 8 - 9 1074 86
AY750170.1 - 8 - 9 1181 86
BC001656.1 - 9 - 10 1338 86

View Full Product Details