The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001173988.1 | NP_001167459.1 | 5 - 6 | 735 | 108 |
NM_001173989.2 | NP_001167460.1 | 5 - 6 | 735 | 108 | |
NM_024718.4 | NP_078994.3 | 5 - 6 | 735 | 108 | |
GenBank mRNA | AF318367.1 | - | 2 - 3 | 2046 | 108 |
AK000963.1 | - | 2 - 3 | 141 | 108 | |
AK001184.1 | - | 4 - 5 | 322 | 108 | |
AK023107.1 | - | 4 - 5 | 329 | 108 | |
AK027586.1 | - | 2 - 3 | 156 | 108 | |
AK094382.1 | - | 5 - 6 | 1058 | 108 | |
AK128154.2 | - | 5 - 6 | 999 | 108 | |
AK307431.1 | - | 6 - 7 | 589 | 108 | |
BC002945.2 | - | 5 - 6 | 498 | 108 | |
BC018960.2 | - | 5 - 6 | 681 | 108 | |
BC021095.1 | - | 5 - 6 | 439 | 108 | |
BC035786.1 | - | 5 - 6 | 690 | 108 | |
DQ099383.1 | - | 2 - 3 | 115 | 108 | |
DQ141240.1 | - | 5 - 6 | 460 | 108 | |
EF156752.1 | - | 5 - 6 | 462 | 108 | |
EF156753.1 | - | 5 - 6 | 461 | 108 | |
GQ169126.1 | - | 5 - 6 | 714 | 108 | |
GQ169127.1 | - | 5 - 6 | 724 | 108 |