The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024896.2 | NP_079172.2 | 13 - 14 | 2440 | 83 |
XM_005251587.1 | XP_005251644.1 | 11 - 12 | 2045 | 83 | |
XM_017015139.1 | XP_016870628.1 | 14 - 15 | 2233 | 83 | |
GenBank mRNA | AB058718.2 | - | 13 - 14 | 2317 | 83 |
AK026962.1 | - | 5 - 6 | 746 | 83 | |
AK093217.1 | - | 11 - 12 | 1616 | 83 | |
AK127218.1 | - | 13 - 14 | 2429 | 83 | |
AL834191.1 | - | 12 - 13 | 2072 | 83 | |
BC031630.1 | - | 5 - 6 | 804 | 83 | |
BC136771.1 | - | 13 - 14 | 2414 | 83 | |
BC136773.1 | - | 13 - 14 | 2414 | 83 |