The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025136.3 | NP_079412.1 | 1 - 2 | 242 | 93 |
GenBank mRNA | AK025840.1 | - | 1 - 2 | 197 | 93 |
BC005059.1 | - | 1 - 2 | 163 | 93 |