The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031955.5 | NP_114161.3 | 3 - 4 | 943 | 61 |
XM_006713778.2 | XP_006713841.1 | 3 - 4 | 917 | 61 | |
XM_017007308.1 | XP_016862797.1 | 3 - 4 | 917 | 61 | |
GenBank mRNA | AF345909.2 | - | 3 - 4 | 889 | 61 |
AF404758.1 | - | 3 - 4 | 889 | 61 | |
AK093041.1 | - | 3 - 4 | 891 | 61 | |
BC034496.2 | - | 3 - 4 | 845 | 61 |