The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032048.2 | NP_114437.2 | 2 - 3 | 416 | 105 |
XM_017026038.1 | XP_016881527.1 | 2 - 3 | 403 | 105 | |
GenBank mRNA | AF270513.1 | - | 2 - 3 | 416 | 105 |
AL552384.3 | - | 2 - 3 | 186 | 105 | |
BC136541.1 | - | 2 - 3 | 370 | 105 |