Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 46857918-46858417 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYL3
Entrez Gene ID
4634
Gene Name
myosin light chain 3
Gene Aliases
CMH8, MLC-lV/sb, MLC1SB, MLC1V, VLC1, VLCl
UniGene
Hs.517939
Gene Chromosome Location
Chr.3: 46857867-46863483 on Build GRCh38
Transcripts
NM_000258.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6164990 c.541G>A p.G181S Chr.3: 46858402-46858402 on Build GRCh38
7425548 c.547A>T p.I183F Chr.3: 46858396-46858396 on Build GRCh38
5665203 c.560-3C>T p.? Chr.3: 46858275-46858275 on Build GRCh38
7378396 c.579G>T p.M193I Chr.3: 46858253-46858253 on Build GRCh38
4117928 c.576C>A p.I192I Chr.3: 46858256-46858256 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757528575 Chr.3: 46857989-46857989 on Build GRCh38
rs751286084 Chr.3: 46858344-46858344 on Build GRCh38
rs750826728 Chr.3: 46858231-46858231 on Build GRCh38
rs963631094 Chr.3: 46858394-46858394 on Build GRCh38
rs758833739 Chr.3: 46858336-46858336 on Build GRCh38
rs1021372893 Chr.3: 46857941-46857941 on Build GRCh38
rs967106687 Chr.3: 46857952-46857952 on Build GRCh38
rs762347889 Chr.3: 46858309-46858309 on Build GRCh38
rs749013158 Chr.3: 46858364-46858364 on Build GRCh38
rs780536817 Chr.3: 46858232-46858232 on Build GRCh38
rs969696050 Chr.3: 46858109-46858109 on Build GRCh38
rs772933222 Chr.3: 46858279-46858279 on Build GRCh38
rs79803923 Chr.3: 46858144-46858144 on Build GRCh38
rs746833285 Chr.3: 46858124-46858124 on Build GRCh38
rs933024341 Chr.3: 46857990-46857990 on Build GRCh38
rs781174287 Chr.3: 46858203-46858203 on Build GRCh38
rs757730888 Chr.3: 46858256-46858257 on Build GRCh38
rs552393482 Chr.3: 46858365-46858365 on Build GRCh38
rs1049678835 Chr.3: 46858220-46858220 on Build GRCh38
rs374329098 Chr.3: 46858386-46858386 on Build GRCh38
rs569290408 Chr.3: 46858305-46858305 on Build GRCh38
rs985833906 Chr.3: 46858367-46858367 on Build GRCh38
rs764865767 Chr.3: 46857975-46857975 on Build GRCh38
rs567490162 Chr.3: 46858074-46858074 on Build GRCh38
rs953115947 Chr.3: 46857942-46857942 on Build GRCh38
rs202234617 Chr.3: 46858235-46858235 on Build GRCh38
rs556971059 Chr.3: 46857966-46857966 on Build GRCh38
rs1042973 Chr.3: 46858026-46858026 on Build GRCh38
rs140829951 Chr.3: 46858391-46858391 on Build GRCh38
rs867195130 Chr.3: 46858216-46858216 on Build GRCh38
rs757691010 Chr.3: 46858219-46858219 on Build GRCh38
rs772146529 Chr.3: 46858286-46858286 on Build GRCh38
rs748154150 Chr.3: 46858241-46858241 on Build GRCh38
rs1012491889 Chr.3: 46857957-46857957 on Build GRCh38
rs764301561 Chr.3: 46858304-46858304 on Build GRCh38
rs1041055773 Chr.3: 46858287-46858287 on Build GRCh38
rs373589725 Chr.3: 46858197-46858197 on Build GRCh38
rs200422816 Chr.3: 46858226-46858226 on Build GRCh38
rs955801208 Chr.3: 46858164-46858164 on Build GRCh38
rs755998566 Chr.3: 46858361-46858361 on Build GRCh38
rs201092907 Chr.3: 46858236-46858236 on Build GRCh38
rs998627417 Chr.3: 46858272-46858272 on Build GRCh38
rs749105574 Chr.3: 46858282-46858282 on Build GRCh38
rs906594778 Chr.3: 46858162-46858162 on Build GRCh38
rs727505197 Chr.3: 46858239-46858239 on Build GRCh38
rs769009588 Chr.3: 46858388-46858388 on Build GRCh38
rs199474710 Chr.3: 46858088-46858088 on Build GRCh38
rs374293766 Chr.3: 46858262-46858262 on Build GRCh38
rs869025487 Chr.3: 46858261-46858261 on Build GRCh38
rs565610572 Chr.3: 46858058-46858058 on Build GRCh38
rs750775381 Chr.3: 46858320-46858320 on Build GRCh38
rs199474709 Chr.3: 46858378-46858378 on Build GRCh38
rs112099443 Chr.3: 46858107-46858107 on Build GRCh38
rs765928342 Chr.3: 46858342-46858342 on Build GRCh38
rs201224325 Chr.3: 46858207-46858207 on Build GRCh38
rs368826045 Chr.3: 46858350-46858350 on Build GRCh38
rs767793908 Chr.3: 46858201-46858201 on Build GRCh38
rs546418537 Chr.3: 46858042-46858042 on Build GRCh38
rs929453165 Chr.3: 46858103-46858103 on Build GRCh38
rs765858022 Chr.3: 46858311-46858311 on Build GRCh38
rs780617884 Chr.3: 46858351-46858351 on Build GRCh38
rs911119001 Chr.3: 46858230-46858230 on Build GRCh38
rs778313994 Chr.3: 46858283-46858285 on Build GRCh38
rs747671640 Chr.3: 46858355-46858355 on Build GRCh38
rs200672141 Chr.3: 46858363-46858363 on Build GRCh38
rs113453656 Chr.3: 46858383-46858383 on Build GRCh38
rs373220421 Chr.3: 46858193-46858193 on Build GRCh38
rs754568155 Chr.3: 46858238-46858238 on Build GRCh38
rs756462246 Chr.3: 46857955-46857955 on Build GRCh38
rs1002240883 Chr.3: 46858188-46858188 on Build GRCh38
rs540912159 Chr.3: 46858214-46858214 on Build GRCh38
rs760655445 Chr.3: 46858302-46858302 on Build GRCh38
rs377399218 Chr.3: 46858204-46858204 on Build GRCh38
rs754150859 Chr.3: 46857977-46857977 on Build GRCh38
rs546968708 Chr.3: 46858003-46858003 on Build GRCh38
rs760847468 Chr.3: 46858205-46858205 on Build GRCh38
rs942237459 Chr.3: 46858038-46858038 on Build GRCh38
rs869025488 Chr.3: 46858275-46858275 on Build GRCh38
rs775829705 Chr.3: 46858296-46858296 on Build GRCh38

View Full Product Details