Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- MEF2C
- Entrez Gene ID
-
4208
- Gene Name
- myocyte enhancer factor 2C
- Gene Aliases
-
C5DELq14.3, DEL5q14.3
- Chromosome Location
-
Chr.5: 88718241 - 88904105 on Build GRCh38
View Full Product Details