The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004575.2 | NP_004566.2 | 1 - 2 | 537 | 57 |
GenBank mRNA | AK314982.1 | - | 1 - 2 | 540 | 57 |
BC136344.1 | - | 1 - 2 | 444 | 57 | |
BC136345.1 | - | 1 - 2 | 444 | 57 | |
BY796838.2 | - | 1 - 2 | 526 | 57 | |
EU439706.1 | - | 1 - 2 | 273 | 57 | |
U06233.1 | - | 1 - 2 | 504 | 57 |