The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021951.2 | NP_068770.2 | 2 - 3 | 689 | 85 |
XM_006716732.1 | XP_006716795.1 | 2 - 3 | 732 | 85 | |
XM_011517770.1 | XP_011516072.1 | 2 - 3 | 625 | 85 | |
XM_011517771.1 | XP_011516073.1 | 2 - 3 | 625 | 85 | |
XM_011517772.1 | XP_011516074.1 | 2 - 3 | 625 | 85 | |
XM_011517773.2 | XP_011516075.1 | 2 - 3 | 444 | 85 | |
GenBank mRNA | AF130728.1 | - | 2 - 3 | 689 | 85 |
AJ276801.1 | - | 2 - 3 | 598 | 85 | |
AK310336.1 | - | 2 - 3 | 427 | 85 | |
AK313594.1 | - | 2 - 3 | 682 | 85 | |
AL162131.1 | - | 2 - 3 | 598 | 85 | |
AY442914.1 | - | 2 - 3 | 689 | 85 | |
BC040847.1 | - | 2 - 3 | 576 | 85 |