The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001225.3 | NP_001216.1 | 2 - 3 | 339 | 113 |
NM_033306.2 | NP_150649.1 | 2 - 3 | 372 | 113 | |
GenBank mRNA | AK057094.1 | - | 2 - 3 | 1177 | 113 |
AK225751.1 | - | 2 - 3 | 114 | ||
AK296081.1 | - | 2 - 3 | 316 | 113 | |
AK304222.1 | - | 2 - 3 | 322 | 113 | |
BC017839.1 | - | 2 - 3 | 305 | 113 | |
CN261701.1 | - | 2 - 3 | 368 | 113 | |
U25804.1 | - | 2 - 3 | 314 | 113 | |
U28014.1 | - | 2 - 3 | 303 | 113 | |
U28976.1 | - | 2 - 3 | 299 | 113 | |
U28977.1 | - | 2 - 3 | 374 | 114 | |
U28978.1 | - | 2 - 3 | 372 | 113 | |
Z48810.1 | - | 2 - 3 | 307 | 113 |