The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012340.4 | NP_036472.2 | 1 - 2 | 349 | 74 |
NM_173091.3 | NP_775114.1 | 1 - 2 | 349 | 74 | |
XM_011528824.2 | XP_011527126.1 | 1 - 2 | 340 | 74 | |
XM_017027851.1 | XP_016883340.1 | 1 - 2 | 340 | 74 | |
GenBank mRNA | BC136418.1 | - | 1 - 2 | 252 | 74 |
BC144074.1 | - | 1 - 2 | 252 | 74 | |
CR993438.1 | - | 1 - 2 | 324 | 74 | |
EU887573.1 | - | 1 - 2 | 129 | 74 | |
EU887578.1 | - | 1 - 2 | 129 | 74 | |
U43341.1 | - | 1 - 2 | 349 | 74 | |
U43342.1 | - | 1 - 2 | 349 | 74 |