Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IVNS1ABP
Entrez Gene ID
10625
Gene Name
influenza virus NS1A binding protein
Gene Aliases
FLARA3, HSPC068, KLHL39, ND1, NS-1, NS1-BP, NS1BP
Chromosome Location
Chr.1: 185292367 - 185317343 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006469.4 NP_006460.2 2 - 3 602 111
XM_005244843.3 XP_005244900.1 2 - 3 558 111
XM_011509079.2 XP_011507381.1 2 - 3 612 111
XM_011509080.2 XP_011507382.1 2 - 3 616 111
XM_017000084.1 XP_016855573.1 2 - 3 558 111
GenBank mRNA AB020657.1 - 2 - 3 609 111
AF161553.1 - 2 - 3 518 111
AF205218.1 - 2 - 3 534 111
AJ012449.1 - 2 - 3 516 111
AK023020.1 - 1 - 2 210 111
AK023123.1 - 2 - 3 546 111
AK292431.1 - 2 - 3 352 111
BC067739.1 - 2 - 3 502 111
DQ443528.1 - 2 - 3 491 111

View Full Product Details