The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159995.2 | NP_001153467.1 | 2 - 3 | 389 | 93 |
NM_001159999.2 | NP_001153471.1 | 2 - 3 | 389 | 93 | |
NM_001160001.2 | NP_001153473.1 | 2 - 3 | 389 | 93 | |
NM_001160002.1 | NP_001153474.1 | 2 - 3 | 796 | 93 | |
NM_001160004.2 | NP_001153476.1 | 2 - 3 | 796 | 93 | |
NM_001160005.1 | NP_001153477.1 | 2 - 3 | 796 | 93 | |
NM_001160007.1 | NP_001153479.1 | 2 - 3 | 796 | 93 | |
NM_001160008.1 | NP_001153480.1 | 2 - 3 | 796 | 93 | |
NM_001322201.1 | NP_001309130.1 | 2 - 3 | 389 | 93 | |
NM_001322202.1 | NP_001309131.1 | 2 - 3 | 389 | 93 | |
NM_004495.3 | NP_004486.2 | 2 - 3 | 796 | 93 | |
NM_013956.4 | NP_039250.2 | 2 - 3 | 796 | 93 | |
NM_013957.4 | NP_039251.2 | 2 - 3 | 796 | 93 | |
NM_013958.3 | NP_039252.2 | 2 - 3 | 796 | 93 | |
NM_013960.4 | NP_039254.1 | 2 - 3 | 796 | 93 | |
NM_013962.2 | NP_039256.2 | 2 - 3 | 1157 | 93 | |
NM_013964.4 | NP_039258.1 | 2 - 3 | 796 | 93 | |
XM_011544512.2 | XP_011542814.1 | 2 - 3 | 579 | 93 | |
XM_017013365.1 | XP_016868854.1 | 2 - 3 | 508 | 93 | |
XM_017013366.1 | XP_016868855.1 | 2 - 3 | 508 | 93 | |
XM_017013367.1 | XP_016868856.1 | 2 - 3 | 391 | 93 | |
XM_017013368.1 | XP_016868857.1 | 2 - 3 | 728 | 93 | |
XM_017013371.1 | XP_016868860.1 | 2 - 3 | 508 | 93 | |
XM_017013372.1 | XP_016868861.1 | 2 - 3 | 508 | 93 | |
GenBank mRNA | AB451331.1 | - | 2 - 3 | 279 | 93 |
AB451475.1 | - | 2 - 3 | 279 | 93 | |
AF026146.1 | - | 2 - 3 | 726 | 93 | |
AK097005.1 | - | 3 - 4 | 643 | 94 | |
AK290730.1 | - | 2 - 3 | 695 | 93 | |
AK298132.1 | - | 2 - 3 | 796 | 93 | |
BC006492.2 | - | 2 - 3 | 768 | 93 | |
BC144446.1 | - | 2 - 3 | 344 | 93 | |
BC150609.1 | - | 2 - 3 | 344 | 93 | |
CN603652.1 | - | 2 - 3 | 307 | 93 | |
CN603654.1 | - | 2 - 3 | 307 | 93 | |
EF372273.1 | - | 2 - 3 | 225 | 93 | |
EF372274.1 | - | 2 - 3 | 225 | 93 | |
EF372275.1 | - | 2 - 3 | 225 | 93 | |
EF372276.1 | - | 2 - 3 | 225 | 93 | |
EF372277.1 | - | 2 - 3 | 225 | 93 | |
EF517295.1 | - | 2 - 3 | 225 | 93 | |
EF517297.1 | - | 2 - 3 | 225 | 93 | |
EU359055.1 | - | 2 - 3 | 276 | 93 | |
EU363508.1 | - | 2 - 3 | 276 | 93 | |
EU363509.1 | - | 2 - 3 | 276 | 93 | |
EU363510.1 | - | 2 - 3 | 276 | 93 | |
GQ983558.1 | - | 2 - 3 | 216 | 93 | |
L12260.1 | - | 2 - 3 | 1185 | 93 | |
L12261.1 | - | 2 - 3 | 672 | 93 | |
M94165.1 | - | 2 - 3 | 411 | 93 | |
M94166.1 | - | 2 - 3 | 371 | 93 | |
M94167.1 | - | 2 - 3 | 739 | 93 | |
M94168.1 | - | 2 - 3 | 771 | 93 | |
S68256.1 | - | 2 - 3 | 351 | 93 | |
U02326.1 | - | 2 - 3 | 387 | 93 |