The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159995.2 | NP_001153467.1 | 6 - 7 | 750 | 65 |
NM_001159999.2 | NP_001153471.1 | 7 - 8 | 801 | 65 | |
NM_001160001.2 | NP_001153473.1 | 5 - 6 | 699 | 65 | |
NM_001322205.1 | NP_001309134.1 | 3 - 4 | 1842 | 65 | |
NM_001322206.1 | NP_001309135.1 | 3 - 4 | 1842 | 65 | |
NM_001322207.1 | NP_001309136.1 | 3 - 4 | 1842 | 65 | |
NM_013956.4 | NP_039250.2 | 7 - 8 | 1208 | 65 | |
XM_006716335.3 | XP_006716398.1 | 3 - 4 | 193 | 65 | |
XM_011544512.2 | XP_011542814.1 | 7 - 8 | 991 | 65 | |
XM_017013367.1 | XP_016868856.1 | 5 - 6 | 701 | 65 | |
XM_017013368.1 | XP_016868857.1 | 5 - 6 | 1038 | 65 | |
XM_017013369.1 | XP_016868858.1 | 3 - 4 | 1810 | 65 | |
GenBank mRNA | AY207002.1 | - | 3 - 4 | 193 | 65 |
CN603661.1 | - | 3 - 4 | 479 | 65 | |
EF372273.1 | - | 5 - 6 | 535 | 65 | |
EF372274.1 | - | 5 - 6 | 535 | 65 | |
EF372276.1 | - | 6 - 7 | 586 | 65 | |
EF517295.1 | - | 5 - 6 | 535 | 65 | |
EF517296.1 | - | 7 - 8 | 637 | 65 | |
EF517297.1 | - | 6 - 7 | 586 | 65 | |
GQ983557.1 | - | 5 - 6 | 699 | 65 | |
M94166.1 | - | 7 - 8 | 783 | 65 | |
U02328.1 | - | 5 - 6 | 414 | 65 |