The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001034116.1 | NP_001029288.1 | 11 - 12 | 1333 | 59 |
NM_001318965.1 | NP_001305894.1 | 10 - 11 | 1292 | 59 | |
NM_001318966.1 | NP_001305895.1 | 11 - 12 | 1383 | 59 | |
NM_001318967.1 | NP_001305896.1 | 11 - 12 | 1326 | 59 | |
NM_001318968.1 | NP_001305897.1 | 10 - 11 | 1222 | 59 | |
NM_001318969.1 | NP_001305898.1 | 11 - 12 | 1307 | 59 | |
NM_015636.3 | NP_056451.3 | 11 - 12 | 1330 | 59 | |
NM_172195.3 | NP_751945.2 | 10 - 11 | 1289 | 59 | |
XM_006712132.1 | XP_006712195.1 | 11 - 12 | 1380 | 59 | |
XM_011533147.2 | XP_011531449.1 | 11 - 12 | 1188 | 59 | |
GenBank mRNA | AF057699.1 | - | 11 - 12 | 1200 | 59 |
AF086520.1 | - | 2 - 3 | 217 | 59 | |
AF112207.1 | - | 11 - 12 | 1209 | 59 | |
AF218009.1 | - | 11 - 12 | 1354 | 59 | |
AJ011305.1 | - | 11 - 12 | 1206 | 59 | |
AJ011306.1 | - | 10 - 11 | 1289 | 59 | |
AL050109.1 | - | 8 - 9 | 803 | 59 | |
BC001870.1 | - | 11 - 12 | 1191 | 59 | |
BC091502.1 | - | 11 - 12 | 1206 | 59 | |
JQ991007.1 | - | 11 - 12 | 1307 | 59 |