The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010983.2 | NP_001010983.1 | 4 - 5 | 1017 | 87 |
NM_001278280.1 | NP_001265209.1 | 4 - 5 | 1013 | 87 | |
NM_001278281.1 | NP_001265210.1 | 4 - 5 | 761 | 87 | |
NM_018446.3 | NP_060916.1 | 3 - 4 | 788 | 87 | |
NM_152932.2 | NP_690909.1 | 3 - 4 | 346 | 87 | |
XM_017006857.1 | XP_016862346.1 | 5 - 6 | 1176 | 87 | |
XM_017006858.1 | XP_016862347.1 | 4 - 5 | 947 | 87 | |
XM_017006859.1 | XP_016862348.1 | 3 - 4 | 496 | 87 | |
GenBank mRNA | AF157318.1 | - | 3 - 4 | 233 | 87 |
AJ271710.1 | - | 2 - 3 | 115 | 87 | |
AK027517.1 | - | 3 - 4 | 87 | ||
AK075031.1 | - | 2 - 3 | 310 | 87 | |
AY358579.1 | - | 3 - 4 | 267 | 87 | |
BC119670.1 | - | 3 - 4 | 232 | 87 | |
BC119671.1 | - | 3 - 4 | 232 | 87 | |
BF725274.1 | - | 4 - 5 | 434 | 87 | |
CR749821.1 | - | 4 - 5 | 470 | 87 |