Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBFOX1
Entrez Gene ID
54715
Gene Name
RNA binding protein, fox-1 homolog 1
Gene Aliases
2BP1, A2BP1, FOX-1, FOX1, HRNBP1
Chromosome Location
Chr.16: 5239749 - 7713343 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142334.1 NP_001135806.1 8 - 9 898 59
NM_001308117.1 NP_001295046.1 10 - 11 1003 59
NM_018723.3 NP_061193.2 10 - 11 1666 59
NM_145891.2 NP_665898.1 7 - 8 990 59
NM_145892.2 NP_665899.1 7 - 8 990 59
NM_145893.2 NP_665900.1 7 - 8 990 59
XM_005255386.3 XP_005255443.1 7 - 8 916 59
XM_005255387.3 XP_005255444.1 7 - 8 895 59
XM_005255388.4 XP_005255445.1 7 - 8 951 59
XM_005255390.3 XP_005255447.1 10 - 11 1788 59
XM_005255391.3 XP_005255448.1 9 - 10 798 59
XM_011522546.2 XP_011520848.1 7 - 8 895 59
XM_011522547.2 XP_011520849.1 7 - 8 895 59
XM_017023318.1 XP_016878807.1 11 - 12 1293 59
XM_017023320.1 XP_016878809.1 10 - 11 1697 59
XM_017023321.1 XP_016878810.1 9 - 10 2759 59
XM_017023322.1 XP_016878811.1 9 - 10 2759 59
XM_017023323.1 XP_016878812.1 7 - 8 916 59
XM_017023324.1 XP_016878813.1 10 - 11 1697 59
XM_017023325.1 XP_016878814.1 8 - 9 1295 59
XM_017023326.1 XP_016878815.1 10 - 11 1788 59
XM_017023327.1 XP_016878816.1 7 - 8 916 59
XM_017023328.1 XP_016878817.1 10 - 11 1697 59
XM_017023330.1 XP_016878819.1 9 - 10 2236 59
XM_017023331.1 XP_016878820.1 7 - 8 2702 59
XM_017023332.1 XP_016878821.1 7 - 8 916 59
XM_017023333.1 XP_016878822.1 9 - 10 798 59
XM_017023335.1 XP_016878824.1 10 - 11 1788 59
XM_017023336.1 XP_016878825.1 9 - 10 798 59
XM_017023339.1 XP_016878828.1 7 - 8 942 59
XM_017023340.1 XP_016878829.1 8 - 9 705 59
GenBank mRNA AB208826.1 - - 1700 59
AF094849.1 - 7 - 8 991 59
AF107203.1 - 10 - 11 1664 59
AF109106.1 - 7 - 8 991 59
AF229057.1 - 7 - 8 993 59
AK001027.1 - 8 - 9 898 59
AK299664.1 - 7 - 8 576 59
AL713700.1 - 9 - 10 1513 59
BC113691.1 - 7 - 8 784 59
KJ534926.1 - 7 - 8 738 59
KJ535000.1 - 7 - 8 738 59

View Full Product Details