Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 145655710-145656235 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MMAA
Entrez Gene ID
166785
Gene Name
methylmalonic aciduria (cobalamin deficiency) cblA type
Gene Aliases
cblA
UniGene
Hs.452864
Gene Chromosome Location
Chr.4: 145619388-145660035 on Build GRCh38
Transcripts
NM_172250.2 XM_011531684.2 XM_011531685.2 XM_011531686.2

SNP Detail

SNP ID Chromosome Location
rs114729477 Chr.4: 145655975-145655975 on Build GRCh38
rs1039741365 Chr.4: 145656017-145656017 on Build GRCh38
rs886059095 Chr.4: 145656023-145656023 on Build GRCh38
rs979866015 Chr.4: 145656201-145656201 on Build GRCh38
rs950628350 Chr.4: 145655925-145655925 on Build GRCh38
rs983793260 Chr.4: 145655926-145655926 on Build GRCh38
rs773103545 Chr.4: 145656041-145656041 on Build GRCh38
rs1024171047 Chr.4: 145656173-145656173 on Build GRCh38
rs72950841 Chr.4: 145655868-145655868 on Build GRCh38
rs934267228 Chr.4: 145656024-145656024 on Build GRCh38
rs565828392 Chr.4: 145656026-145656026 on Build GRCh38
rs530774368 Chr.4: 145655774-145655774 on Build GRCh38
rs533580327 Chr.4: 145656004-145656004 on Build GRCh38
rs936007824 Chr.4: 145656015-145656015 on Build GRCh38
rs945870579 Chr.4: 145655962-145655962 on Build GRCh38
rs1034155575 Chr.4: 145656202-145656202 on Build GRCh38
rs886059093 Chr.4: 145655776-145655776 on Build GRCh38
rs1015090391 Chr.4: 145656040-145656040 on Build GRCh38
rs942837582 Chr.4: 145656029-145656029 on Build GRCh38
rs886059094 Chr.4: 145655974-145655974 on Build GRCh38
rs1005070010 Chr.4: 145655769-145655769 on Build GRCh38
rs1034056790 Chr.4: 145656058-145656058 on Build GRCh38
rs994989620 Chr.4: 145656057-145656057 on Build GRCh38
rs891412293 Chr.4: 145656028-145656028 on Build GRCh38
rs992743532 Chr.4: 145655931-145655931 on Build GRCh38
rs917113255 Chr.4: 145655947-145655947 on Build GRCh38
rs899840258 Chr.4: 145656023-145656023 on Build GRCh38
rs112882351 Chr.4: 145656034-145656034 on Build GRCh38
rs973053760 Chr.4: 145655822-145655822 on Build GRCh38
rs979942593 Chr.4: 145655991-145655991 on Build GRCh38
rs374413811 Chr.4: 145656108-145656108 on Build GRCh38
rs376383308 Chr.4: 145655814-145655814 on Build GRCh38
rs548243828 Chr.4: 145656137-145656137 on Build GRCh38
rs1026066167 Chr.4: 145655898-145655898 on Build GRCh38
rs181600698 Chr.4: 145656014-145656014 on Build GRCh38
rs1024252024 Chr.4: 145655966-145655966 on Build GRCh38
rs967617277 Chr.4: 145656176-145656176 on Build GRCh38
rs958648520 Chr.4: 145655869-145655869 on Build GRCh38
rs958343240 Chr.4: 145656064-145656064 on Build GRCh38
rs995515398 Chr.4: 145655877-145655877 on Build GRCh38
rs74409983 Chr.4: 145655740-145655740 on Build GRCh38
rs749692437 Chr.4: 145656093-145656093 on Build GRCh38
rs887258086 Chr.4: 145655765-145655765 on Build GRCh38
rs142549499 Chr.4: 145656166-145656166 on Build GRCh38
rs888072942 Chr.4: 145655776-145655776 on Build GRCh38
rs796780340 Chr.4: 145656019-145656019 on Build GRCh38
rs887290820 Chr.4: 145656032-145656032 on Build GRCh38
rs536872965 Chr.4: 145655837-145655837 on Build GRCh38
rs957116242 Chr.4: 145656204-145656204 on Build GRCh38

View Full Product Details