The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013397.5 | NP_037529.3 | 2 - 3 | 233 | 66 |
GenBank mRNA | BC110458.1 | - | 1 - 2 | 160 | 66 |
BC110459.1 | - | 1 - 2 | 160 | 66 | |
DA875485.1 | - | 2 - 3 | 210 | 66 | |
DA957684.1 | - | 2 - 3 | 233 | 66 |