The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022841.5 | NP_073752.5 | 9 - 9 | 3444 | 85 |
XM_005254603.3 | XP_005254660.2 | 10 - 10 | 3538 | 85 | |
XM_011521925.2 | XP_011520227.1 | 9 - 9 | 3376 | 85 | |
XM_017022506.1 | XP_016877995.1 | 10 - 10 | 6261 | 85 | |
XM_017022507.1 | XP_016877996.1 | 9 - 9 | 3504 | 85 | |
XM_017022508.1 | XP_016877997.1 | 8 - 8 | 4673 | 85 | |
GenBank mRNA | AK023056.1 | - | 1 - 1 | 1739 | 85 |
AK126103.1 | - | 2 - 2 | 2596 | 85 | |
AK128045.1 | - | 9 - 9 | 3381 | 85 | |
AL832584.1 | - | 1 - 1 | 2254 | 85 | |
AL833367.1 | - | 1 - 1 | 1671 | 85 | |
AL834302.1 | - | 1 - 1 | 253 | 85 | |
BC112936.1 | - | 7 - 7 | 3264 | 85 | |
BC143438.1 | - | 7 - 7 | 3201 | 85 |