Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARMC10
Entrez Gene ID
83787
Gene Name
armadillo repeat containing 10
Gene Aliases
PNAS-112, PNAS112, PSEC0198, SVH
Chromosome Location
Chr.7: 103074727 - 103099763 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161009.2 NP_001154481.1 5 - 6 1065 69
NM_001161010.2 NP_001154482.1 5 - 6 993 69
NM_001161012.2 NP_001154484.1 4 - 5 888 69
NM_031905.4 NP_114111.2 6 - 7 1170 69
XM_011516601.2 XP_011514903.1 5 - 6 1243 69
XM_011516602.2 XP_011514904.1 5 - 6 604 69
XM_017012684.1 XP_016868173.1 7 - 8 1351 69
XM_017012686.1 XP_016868175.1 6 - 7 1174 69
XM_017012687.1 XP_016868176.1 5 - 6 583 69
GenBank mRNA AK075500.1 - 6 - 7 911 69
AK095169.1 - 1 - 2 1623 69
AK291818.1 - 5 - 6 793 69
AY150851.1 - 4 - 5 888 69
AY150852.1 - 5 - 6 993 69
AY150853.1 - 5 - 6 1065 69
AY150854.1 - 6 - 7 1170 69
BC003586.1 - 5 - 6 785 69

View Full Product Details