Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC37A4
Entrez Gene ID
2542
Gene Name
solute carrier family 37 member 4
Gene Aliases
G6PT1, G6PT2, G6PT3, GSD1b, GSD1c, GSD1d, PRO0685, TRG-19, TRG19
Chromosome Location
Chr.11: 119024351 - 119030906 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164277.1 NP_001157749.1 3 - 4 903 141
NM_001164278.1 NP_001157750.1 3 - 4 587 141
NM_001164280.1 NP_001157752.1 1 - 2 345 141
NM_001467.5 NP_001458.1 2 - 3 399 141
GenBank mRNA AF110819.1 - 1 - 2 146 141
AF110820.1 - 1 - 2 146 141
AF110821.1 - 1 - 2 146 141
AF110822.1 - 1 - 2 146 141
AK289642.1 - 3 - 4 871 141
AK298395.1 - 3 - 4 283 141
AK312628.1 - 3 - 4 569 141
AY423732.1 - 1 - 2 166 141
BC002400.2 - 1 - 2 347 141
BC003589.1 - 1 - 2 347 141
BC014663.1 - 1 - 2 354 141
BC015650.2 - 2 - 3 819 141
BC064563.1 - 1 - 2 342 141
CR456733.1 - 1 - 2 146 141
DA485567.1 - 2 - 3 373 141
DC295438.1 - 2 - 3 399 141
Y15409.1 - 1 - 2 315 141

View Full Product Details