The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032367.2 | NP_115743.1 | 1 - 2 | 95 | 62 |
GenBank mRNA | AK223624.1 | - | 1 - 2 | 109 | 62 |
BC007239.1 | - | 1 - 2 | 95 | 62 |