The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321431.1 | NP_001308360.1 | 30 - 31 | 3981 | 73 |
NM_032447.4 | NP_115823.3 | 30 - 31 | 4256 | 73 | |
XM_017027372.1 | XP_016882861.1 | 30 - 31 | 3980 | 73 | |
XM_017027373.1 | XP_016882862.1 | 30 - 31 | 4239 | 73 | |
XM_017027374.1 | XP_016882863.1 | 29 - 30 | 4143 | 73 | |
XM_017027375.1 | XP_016882864.1 | 30 - 31 | 4239 | 73 | |
XM_017027376.1 | XP_016882865.1 | 30 - 31 | 4239 | 73 | |
XM_017027377.1 | XP_016882866.1 | 11 - 12 | 1306 | 73 | |
XM_017027378.1 | XP_016882867.1 | 3 - 4 | 533 | 73 | |
XM_017027379.1 | XP_016882868.1 | 31 - 32 | 4365 | 73 | |
GenBank mRNA | AB053450.2 | - | 29 - 30 | 3862 | 73 |
AY165863.1 | - | 30 - 31 | 4256 | 73 | |
AY165864.1 | - | 30 - 31 | 3981 | 73 | |
AY165865.1 | - | 29 - 30 | 4127 | 73 | |
BC153882.1 | - | 29 - 30 | 3862 | 73 | |
CN369190.1 | - | 2 - 3 | 200 | 73 |