Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 78020565-78021111 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP3B1
Entrez Gene ID
8546
Gene Name
adaptor related protein complex 3 beta 1 subunit
Gene Aliases
ADTB3, ADTB3A, HPS, HPS2, PE
UniGene
Hs.532091
Gene Chromosome Location
Chr.5: 78002326-78294755 on Build GRCh38
Transcripts
NM_001271769.1 NM_003664.4 XM_005248618.3 XM_017010001.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7297920 c.2899A>G p.K967E Chr.5: 78020785-78020785 on Build GRCh38
3715265 c.2941G>A p.E981K Chr.5: 78020743-78020743 on Build GRCh38
4662251 c.2896A>G p.T966A Chr.5: 78020788-78020788 on Build GRCh38
6033582 c.2960C>A p.A987D Chr.5: 78020724-78020724 on Build GRCh38
3366165 c.2926C>T p.Q976* Chr.5: 78020758-78020758 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs927065568 Chr.5: 78021055-78021055 on Build GRCh38
rs773877723 Chr.5: 78020766-78020766 on Build GRCh38
rs17190102 Chr.5: 78020999-78020999 on Build GRCh38
rs762148185 Chr.5: 78020645-78020645 on Build GRCh38
rs929892456 Chr.5: 78020844-78020844 on Build GRCh38
rs1028629462 Chr.5: 78021051-78021051 on Build GRCh38
rs191976809 Chr.5: 78020894-78020894 on Build GRCh38
rs183954514 Chr.5: 78020982-78020982 on Build GRCh38
rs117001247 Chr.5: 78020994-78020994 on Build GRCh38
rs527847744 Chr.5: 78020651-78020651 on Build GRCh38
rs541639415 Chr.5: 78020977-78020977 on Build GRCh38
rs187393876 Chr.5: 78020632-78020632 on Build GRCh38
rs761000168 Chr.5: 78020781-78020781 on Build GRCh38
rs779085657 Chr.5: 78020808-78020808 on Build GRCh38
rs989506778 Chr.5: 78020923-78020923 on Build GRCh38
rs780150041 Chr.5: 78020664-78020664 on Build GRCh38
rs753937992 Chr.5: 78020666-78020666 on Build GRCh38
rs755171790 Chr.5: 78020672-78020672 on Build GRCh38
rs747919557 Chr.5: 78020891-78020891 on Build GRCh38
rs956203484 Chr.5: 78020991-78020991 on Build GRCh38
rs755086014 Chr.5: 78020805-78020805 on Build GRCh38
rs752854757 Chr.5: 78020818-78020818 on Build GRCh38
rs770490934 Chr.5: 78021016-78021016 on Build GRCh38
rs766616397 Chr.5: 78020784-78020784 on Build GRCh38
rs752101910 Chr.5: 78020641-78020641 on Build GRCh38
rs924224675 Chr.5: 78020893-78020893 on Build GRCh38
rs541478386 Chr.5: 78021002-78021002 on Build GRCh38
rs375250754 Chr.5: 78020682-78020682 on Build GRCh38
rs886060770 Chr.5: 78020778-78020778 on Build GRCh38
rs576731963 Chr.5: 78020609-78020609 on Build GRCh38
rs769476549 Chr.5: 78020738-78020738 on Build GRCh38
rs994690906 Chr.5: 78021021-78021021 on Build GRCh38
rs775073514 Chr.5: 78020742-78020742 on Build GRCh38
rs111490821 Chr.5: 78020889-78020889 on Build GRCh38
rs554498493 Chr.5: 78020929-78020929 on Build GRCh38
rs750787338 Chr.5: 78020652-78020652 on Build GRCh38
rs888748595 Chr.5: 78021054-78021054 on Build GRCh38
rs776144735 Chr.5: 78021024-78021024 on Build GRCh38
rs369459422 Chr.5: 78020787-78020787 on Build GRCh38
rs754043750 Chr.5: 78020797-78020797 on Build GRCh38
rs149212770 Chr.5: 78020773-78020773 on Build GRCh38
rs200516181 Chr.5: 78020658-78020658 on Build GRCh38
rs779017901 Chr.5: 78020689-78020689 on Build GRCh38
rs745653147 Chr.5: 78020737-78020737 on Build GRCh38
rs906323243 Chr.5: 78020595-78020595 on Build GRCh38
rs746992047 Chr.5: 78020697-78020697 on Build GRCh38
rs149754962 Chr.5: 78020832-78020832 on Build GRCh38
rs570567595 Chr.5: 78020681-78020681 on Build GRCh38
rs762495152 Chr.5: 78020750-78020750 on Build GRCh38
rs1039124493 Chr.5: 78020939-78020939 on Build GRCh38
rs768204199 Chr.5: 78020763-78020763 on Build GRCh38
rs745672418 Chr.5: 78020826-78020826 on Build GRCh38
rs757192302 Chr.5: 78020819-78020819 on Build GRCh38
rs887793843 Chr.5: 78020896-78020896 on Build GRCh38
rs1004802352 Chr.5: 78020623-78020623 on Build GRCh38
rs145450651 Chr.5: 78020898-78020898 on Build GRCh38
rs139968311 Chr.5: 78020769-78020769 on Build GRCh38
rs771013399 Chr.5: 78020711-78020711 on Build GRCh38
rs1015294748 Chr.5: 78020629-78020629 on Build GRCh38
rs1019788737 Chr.5: 78020785-78020785 on Build GRCh38
rs769518072 Chr.5: 78020830-78020830 on Build GRCh38
rs948124240 Chr.5: 78020963-78020963 on Build GRCh38
rs762271046 Chr.5: 78020767-78020767 on Build GRCh38
rs575858293 Chr.5: 78020825-78020828 on Build GRCh38
rs763689120 Chr.5: 78020945-78020948 on Build GRCh38
rs959782116 Chr.5: 78021061-78021061 on Build GRCh38
rs772984876 Chr.5: 78020984-78020984 on Build GRCh38
rs150453581 Chr.5: 78020724-78020724 on Build GRCh38

View Full Product Details