The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033952.2 | NP_001029124.1 | 3 - 4 | 308 | 65 |
NM_001741.2 | NP_001732.1 | 3 - 4 | 284 | 65 | |
NR_125898.1 | 3 - 4 | 284 | 65 | ||
XM_017018283.1 | XP_016873772.1 | 3 - 4 | 359 | 65 | |
XM_017018284.1 | XP_016873773.1 | 3 - 4 | 383 | 65 | |
GenBank mRNA | BC069684.1 | - | 3 - 4 | 284 | 65 |
BC069704.1 | - | 3 - 4 | 284 | 65 | |
BC069760.1 | - | 3 - 4 | 284 | 65 | |
BC069778.1 | - | 3 - 4 | 284 | 65 | |
BC093753.1 | - | 3 - 4 | 295 | 65 | |
BC101599.1 | - | 3 - 4 | 271 | 65 | |
BC143562.1 | - | 3 - 4 | 295 | 65 | |
M26095.1 | - | 3 - 4 | 267 | 65 | |
M64486.1 | - | 3 - 4 | 272 | 65 | |
X00356.1 | - | 3 - 4 | 303 | 65 | |
X02330.1 | - | 1 - 2 | 89 | 65 | |
X03662.1 | - | 3 - 4 | 267 | 65 |