The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001872.4 | NP_001863.3 | 6 - 7 | 662 | 79 |
XM_017020393.1 | XP_016875882.1 | 6 - 7 | 605 | 79 | |
GenBank mRNA | AK290829.1 | - | 6 - 7 | 610 | 79 |
BC007057.1 | - | 6 - 7 | 619 | 79 | |
BT006936.1 | - | 6 - 7 | 595 | 79 | |
M75106.1 | - | 6 - 7 | 614 | 79 |