Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 148991403-148991913 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SH3TC2
Entrez Gene ID
79628
Gene Name
SH3 domain and tetratricopeptide repeats 2
Gene Aliases
CMT4C, MNMN
UniGene
Hs.483784
Gene Chromosome Location
Chr.5: 148982150-149063174 on Build GRCh38
Transcripts
NM_024577.3

SNP Detail

SNP ID Chromosome Location
rs546309709 Chr.5: 148991609-148991609 on Build GRCh38
rs1034139567 Chr.5: 148991528-148991528 on Build GRCh38
rs141603977 Chr.5: 148991507-148991507 on Build GRCh38
rs539673321 Chr.5: 148991560-148991560 on Build GRCh38
rs893538035 Chr.5: 148991491-148991491 on Build GRCh38
rs961759889 Chr.5: 148991712-148991712 on Build GRCh38
rs906820258 Chr.5: 148991578-148991578 on Build GRCh38
rs780513309 Chr.5: 148991717-148991717 on Build GRCh38
rs138600983 Chr.5: 148991481-148991481 on Build GRCh38
rs1006102270 Chr.5: 148991657-148991657 on Build GRCh38
rs187321144 Chr.5: 148991584-148991584 on Build GRCh38
rs922586072 Chr.5: 148991765-148991765 on Build GRCh38
rs555058516 Chr.5: 148991807-148991807 on Build GRCh38
rs76955068 Chr.5: 148991742-148991742 on Build GRCh38
rs573227616 Chr.5: 148991853-148991853 on Build GRCh38
rs182516724 Chr.5: 148991871-148991871 on Build GRCh38
rs1019905533 Chr.5: 148991863-148991863 on Build GRCh38
rs910426200 Chr.5: 148991828-148991828 on Build GRCh38
rs1010340871 Chr.5: 148991857-148991857 on Build GRCh38
rs534278887 Chr.5: 148991864-148991864 on Build GRCh38
rs536015698 Chr.5: 148991769-148991769 on Build GRCh38
rs943250289 Chr.5: 148991839-148991839 on Build GRCh38
rs1041211262 Chr.5: 148991784-148991784 on Build GRCh38
rs991317174 Chr.5: 148991563-148991563 on Build GRCh38
rs949425932 Chr.5: 148991684-148991684 on Build GRCh38
rs571249769 Chr.5: 148991641-148991641 on Build GRCh38
rs771420694 Chr.5: 148991574-148991574 on Build GRCh38
rs190671404 Chr.5: 148991766-148991766 on Build GRCh38
rs1020556575 Chr.5: 148991533-148991533 on Build GRCh38
rs931549453 Chr.5: 148991741-148991741 on Build GRCh38
rs183392891 Chr.5: 148991440-148991440 on Build GRCh38
rs369930982 Chr.5: 148991739-148991739 on Build GRCh38
rs1019325107 Chr.5: 148991429-148991429 on Build GRCh38
rs769382913 Chr.5: 148991674-148991674 on Build GRCh38
rs371274790 Chr.5: 148991876-148991876 on Build GRCh38
rs375496558 Chr.5: 148991734-148991734 on Build GRCh38
rs1009877717 Chr.5: 148991449-148991449 on Build GRCh38
rs544508111 Chr.5: 148991889-148991889 on Build GRCh38
rs576816749 Chr.5: 148991885-148991885 on Build GRCh38
rs958533318 Chr.5: 148991546-148991546 on Build GRCh38

View Full Product Details