Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DTX1
Entrez Gene ID
1840
Gene Name
deltex E3 ubiquitin ligase 1
Gene Aliases
hDx-1
Chromosome Location
Chr.12: 113056711 - 113098028 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004416.2 NP_004407.2 1 - 2 595 110
XM_011538009.2 XP_011536311.1 2 - 3 1237 110
GenBank mRNA AF053700.1 - 1 - 2 762 110
BC048216.1 - 1 - 2 537 110

View Full Product Details