The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320227.1 | NP_001307156.1 | 4 - 5 | 417 | 80 |
NM_015938.4 | NP_057022.2 | 4 - 5 | 417 | 80 | |
XM_005247511.1 | XP_005247568.1 | 4 - 5 | 367 | 80 | |
XM_005247512.1 | XP_005247569.1 | 4 - 5 | 698 | 80 | |
GenBank mRNA | AF132941.1 | - | 4 - 5 | 388 | 80 |
AK225664.1 | - | 4 - 5 | 399 | 80 | |
AK308865.1 | - | 4 - 5 | 396 | 80 | |
BC013317.1 | - | 4 - 5 | 400 | 80 |