Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FN1
Entrez Gene ID
2335
Gene Name
fibronectin 1
Gene Aliases
CIG, ED-B, FINC, FN, FNZ, GFND, GFND2, LETS, MSF
Chromosome Location
Chr.2: 215360440 - 215436167 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306129.1 NP_001293058.1 20 - 21 3525 78
NM_001306130.1 NP_001293059.1 20 - 21 3525 78
NM_001306131.1 NP_001293060.1 20 - 21 3525 78
NM_001306132.1 NP_001293061.1 20 - 21 3525 78
NM_002026.3 NP_002017.1 20 - 21 3523 78
NM_212474.2 NP_997639.1 20 - 21 3523 78
NM_212476.2 NP_997641.1 20 - 21 3523 78
NM_212478.2 NP_997643.1 20 - 21 3523 78
NM_212482.2 NP_997647.1 20 - 21 3523 78
XM_005246397.1 XP_005246454.1 20 - 21 3622 78
XM_005246398.1 XP_005246455.1 20 - 21 3622 78
XM_005246399.1 XP_005246456.1 20 - 21 3622 78
XM_005246401.1 XP_005246458.1 20 - 21 3622 78
XM_005246402.1 XP_005246459.1 20 - 21 3622 78
XM_005246403.1 XP_005246460.1 20 - 21 3622 78
XM_005246404.1 XP_005246461.1 20 - 21 3622 78
XM_005246405.1 XP_005246462.1 20 - 21 3622 78
XM_005246406.1 XP_005246463.1 20 - 21 3622 78
XM_005246407.1 XP_005246464.1 20 - 21 3622 78
XM_005246408.1 XP_005246465.1 20 - 21 3622 78
XM_005246409.1 XP_005246466.1 20 - 21 3622 78
XM_005246410.1 XP_005246467.1 20 - 21 3622 78
XM_005246411.1 XP_005246468.1 20 - 21 3622 78
XM_005246412.1 XP_005246469.1 20 - 21 3622 78
XM_005246414.1 XP_005246471.1 20 - 21 3622 78
XM_005246416.1 XP_005246473.1 20 - 21 3622 78
XM_017003692.1 XP_016859181.1 20 - 21 3622 78
XM_017003693.1 XP_016859182.1 20 - 21 3622 78
XM_017003694.1 XP_016859183.1 20 - 21 3622 78
XM_017003695.1 XP_016859184.1 20 - 21 3622 78
GenBank mRNA AB191261.1 - 20 - 21 3523 78
AB209840.1 - 20 - 21 3515 78
AL832202.1 - 20 - 21 3524 78
AL832771.1 - 20 - 21 3524 78
BC117176.1 - 20 - 21 3503 78
BC143754.1 - 20 - 21 3503 78
BC143763.1 - 20 - 21 3503 78
BX537590.1 - 20 - 21 3524 78
BX538017.1 - 20 - 21 3524 78
BX538018.1 - 20 - 21 3524 78
BX640608.1 - 20 - 21 3523 78
BX640731.1 - 20 - 21 3524 78
BX640802.1 - 20 - 21 3525 78
BX640803.1 - 20 - 21 3523 78
BX640875.1 - 20 - 21 3524 78
BX640999.1 - 15 - 16 2499 78
BX649182.1 - 20 - 21 3525 78
CR749281.1 - 20 - 21 3562 78
CR749316.1 - 20 - 21 3522 78
CR749317.1 - 20 - 21 3522 78
X02761.1 - 20 - 21 3177 78

View Full Product Details