Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155237491-155238002 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBA
Entrez Gene ID
2629
Gene Name
glucosylceramidase beta
Gene Aliases
GBA1, GCB, GLUC
UniGene
Hs.282997
Gene Chromosome Location
Chr.1: 155234448-155244862 on Build GRCh38
Transcripts
NM_000157.3 NM_001005741.2 NM_001005742.2 NM_001171811.1 NM_001171812.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5992711 c.773C>G p.A258G Chr.1: 155237567-155237567 on Build GRCh38
463093 c.837G>A p.L279L Chr.1: 155237503-155237503 on Build GRCh38
7420495 c.832G>A p.G278R Chr.1: 155237508-155237508 on Build GRCh38
1472710 c.840G>C p.L280F Chr.1: 155237500-155237500 on Build GRCh38
2210243 c.772G>T p.A258S Chr.1: 155237568-155237568 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377217353 Chr.1: 155237623-155237623 on Build GRCh38
rs1017286953 Chr.1: 155237674-155237674 on Build GRCh38
rs776807095 Chr.1: 155237606-155237606 on Build GRCh38
rs28678003 Chr.1: 155237942-155237942 on Build GRCh38
rs900737195 Chr.1: 155237876-155237876 on Build GRCh38
rs773893138 Chr.1: 155237767-155237767 on Build GRCh38
rs549565365 Chr.1: 155237914-155237914 on Build GRCh38
rs121908303 Chr.1: 155237577-155237577 on Build GRCh38
rs992159581 Chr.1: 155237800-155237800 on Build GRCh38
rs529758518 Chr.1: 155237676-155237676 on Build GRCh38
rs780805088 Chr.1: 155237655-155237655 on Build GRCh38
rs772063113 Chr.1: 155237597-155237597 on Build GRCh38
rs372809157 Chr.1: 155237616-155237616 on Build GRCh38
rs111777259 Chr.1: 155237858-155237858 on Build GRCh38
rs927225220 Chr.1: 155237788-155237788 on Build GRCh38
rs929351376 Chr.1: 155237807-155237807 on Build GRCh38
rs548230876 Chr.1: 155237708-155237708 on Build GRCh38
rs74500255 Chr.1: 155237576-155237576 on Build GRCh38
rs938033488 Chr.1: 155237789-155237789 on Build GRCh38
rs932763712 Chr.1: 155237927-155237927 on Build GRCh38
rs140335079 Chr.1: 155237596-155237596 on Build GRCh38
rs778092646 Chr.1: 155237542-155237543 on Build GRCh38
rs759909325 Chr.1: 155237627-155237627 on Build GRCh38
rs936299790 Chr.1: 155237568-155237568 on Build GRCh38
rs1047926700 Chr.1: 155237831-155237831 on Build GRCh38
rs755722145 Chr.1: 155237557-155237557 on Build GRCh38
rs770615819 Chr.1: 155237572-155237572 on Build GRCh38
rs562620247 Chr.1: 155237667-155237667 on Build GRCh38
rs979955939 Chr.1: 155237774-155237774 on Build GRCh38
rs368734596 Chr.1: 155237607-155237607 on Build GRCh38
rs1021392287 Chr.1: 155237751-155237751 on Build GRCh38
rs891559010 Chr.1: 155237976-155237976 on Build GRCh38
rs546199839 Chr.1: 155237560-155237560 on Build GRCh38
rs763295309 Chr.1: 155237618-155237618 on Build GRCh38
rs997191555 Chr.1: 155237695-155237695 on Build GRCh38
rs900197682 Chr.1: 155237677-155237677 on Build GRCh38
rs552220439 Chr.1: 155237775-155237775 on Build GRCh38
rs532811497 Chr.1: 155237730-155237730 on Build GRCh38
rs770035116 Chr.1: 155237609-155237609 on Build GRCh38
rs990173350 Chr.1: 155237903-155237903 on Build GRCh38
rs918007446 Chr.1: 155237801-155237801 on Build GRCh38
rs777370349 Chr.1: 155237561-155237561 on Build GRCh38
rs555279454 Chr.1: 155237708-155237708 on Build GRCh38
rs367840697 Chr.1: 155237625-155237625 on Build GRCh38
rs537692653 Chr.1: 155237890-155237890 on Build GRCh38
rs572370038 Chr.1: 155237545-155237545 on Build GRCh38
rs955804501 Chr.1: 155237723-155237723 on Build GRCh38
rs1039730395 Chr.1: 155237862-155237862 on Build GRCh38
rs1006437355 Chr.1: 155237658-155237658 on Build GRCh38
rs752226690 Chr.1: 155237552-155237552 on Build GRCh38
rs376957951 Chr.1: 155237903-155237904 on Build GRCh38
rs749014188 Chr.1: 155237564-155237564 on Build GRCh38
rs2009578 Chr.1: 155237835-155237835 on Build GRCh38
rs76725886 Chr.1: 155237531-155237531 on Build GRCh38
rs762488 Chr.1: 155237758-155237758 on Build GRCh38
rs888046581 Chr.1: 155237644-155237644 on Build GRCh38
rs866077794 Chr.1: 155237859-155237859 on Build GRCh38

View Full Product Details